missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls

Product Code. 15259804 Shop All Cytiva Products
Change view
Click to view available options
Width (Metric):
15 cm
20 cm
30 cm
Pore Size:
0.1 μm
0.2 μm
0.45 μm
Unit Size:
1 roll
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Pore Size Unit Size
15259804 15 cm 0.45 μm 1 roll
15239794 30 cm 0.1 μm 1 roll
15259794 30 cm 0.45 μm 1 roll
15249794 30 cm 0.2 μm 1 roll
15289794 20 cm 0.1 μm 1 roll
15299794 20 cm 0.2 μm 1 roll
15239804 15 cm 0.1 μm 1 roll
15209804 20 cm 0.45 μm 1 roll
15249804 15 cm 0.2 μm 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 15259804 Supplier Cytiva Supplier No. 10600012
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use this versatile nitrocellulose membrane for excellent binding affinity for small proteins and peptides, as well as nucleic acids. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.

Packed in rolls.

  • 100% pure nitrocellulose
  • High affinity for proteins
  • Blocking ability
  • Compatible with a range of detection methods, including chemiluminescent, chromogenic, and fluorescent

TRUSTED_SUSTAINABILITY

Tekniske data

Length (Metric) 4 m
Width (Metric) 15 cm
For Use With (Application) Western blots, dot-blot assays, and other protein or nucleic acid methods
Format Roll
Material 100% Pure Nitrocellulose
Pore Size 0.45 μm
compliance-icons
Produktidentifikator
  • Amersham Protran 0.45
Signalord
  • Warning
Farekategori
  • Flammable solid Category 2
Faresætning
  • H228-Flammable solid.
Sikkerhedssætning
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.