missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Protran™ Premium NC Nitrocellulose Western Blotting Sandwiches

Product Code. 15269884 Shop All Cytiva Products
Change view
Click to view available options
Length (Metric):
8 cm
Width (Metric):
9 cm
Pore Size:
0.2 μm
0.45 μm
Unit Size:
Pack of 10
Product Code. Length (Metric) Width (Metric) Pore Size unitSize
15269884 8 cm 9 cm 0.2 μm Pack of 10
15229884 8 cm 9 cm 0.45 μm Pack of 10
2 options
This item is not returnable. View return policy

Product Code. 15269884

Brand: Cytiva 10600118

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Precut, preassembled Protran Premium NC Nitrocellulose membrane and filter paper sandwiches save time

  • Preassembled with two Whatman™ medium thickness 3MM Chr (0.34mm) filter papers to simplify set up
  • Fit most precase gel formats

TRUSTED_SUSTAINABILITY

Specifications

Length (Metric) 8 cm
Width (Metric) 9 cm
Quantity Pack of 10
Format Sandwich
Pore Size 0.2 μm
compliance-icons
Product Identifier
  • amersham protran supported 0.2µm NC 200mm x 200mm
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.