missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ TE 77 Semi-Dry Transfer Unit

Product Code. 10382910 Shop All Cytiva Products
Change view
Click to view available options
Gel Dimensions:
21 x 26 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Gel Dimensions unitSize
10382910 21 x 26 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10382910 Supplier Cytiva Supplier No. TE77

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

 

  • Long-lasting, perforated platinum-titanium and stainless-steel electrodes give uniform, contamination-free transfers
  • Requires minimal buffer, only enough to saturate blotting papers and membranes
  • Uses low current (0.5A) and voltage (30V) for transfers: safer and more economical
  • Suitable for gels up to 21 × 26cm
  • Tested for use with Amersham™ ECL Western Blotting reagents
  • Unique automatic stopping feature; transfer is stopped when buffer gets depleted, preventing overheating

 

TRUSTED_SUSTAINABILITY

Specifications

Description ECL Semi-Dry Blotters are designed to deliver outstanding electrophoretic transfer results using low current and voltage with a minimal amount of buffer. Units are available with and without built-in power supplies to give maximum flexibility.
Type Semi-Dry Transfer Unit
Gel Dimensions 21 x 26 cm
Includes 25 sheets blotting paper, 50 sheets cellophane, solid mask and mask for 14 x 16 cm gels
Model TE 77
Width (Metric) 38 cm
Height (Metric) 9 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.