missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ AMG 18 hydrochloride

Product Code. 18728248
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18728248 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18728248 Supplier R&D Systems™ Supplier No. 6166/5
  • ONLINE EXCLUSIVE PRICE
    408.22 €
    367.50 € / 5mg
    Save 40.72 €
    9% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. IRE1 Inhibitors

AMG 18 hydrochloride is a potent and selective IRE1α inhibitor (IC50 values are 13 and 99 nM for recombinant and cellular IRE1α, respectively). Exhibits selectivity for IRE1α in a panel of over 100 other kinases.
TRUSTED_SUSTAINABILITY

Specifications

CAS 2250019-92-0
Synonym 2-Chloro-N-[6-methyl-5-[[3-[2-[(3S)-3-piperidinylamino]-4-pyrimidinyl]-2-pyridinyl]oxy]-1-naphthalenyl]benzenesulfonamide hydrochloride
Quantity 5 mg
Product Line Tocris
Molecular Formula C31H29ClN6O3S.HCl
Formula Weight 637.58
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.