missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AMG 510
GREENER_CHOICE

Product Code. 18717466
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18717466

Brand: Tocris Bioscience™ 7713/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective KRASG12C inhibitor

AMG 510 is a potent and selective covalent KRASG12C inhibitor (IC50 = 90 nM). Also a potent inhibitor of phospho-ERK (IC50 = 68 nM). In an in vivo model of non-small cell lung cancer, AMG 510 causes regression of tumors, an increase in T-cell recruitment and a pro-inflammatory tumor microenvironment. Synergises with MEK inhibitors and Carboplatin (Cat. No. 2626) and is orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1 R)-6-Fluoro-7-(2-fluoro-6-hydroxyphenyl)-1-[4-methyl-2-(1-methylethyl)-3-pyridinyl]-4-[(2 S)-2-methyl-4-(1-oxo-2-propen-1-yl)-1-piperazinyl]pyrido[2,3-d]pyrimidin-2(1 H)-one
CAS 2252403-56-6
Quantity 10 mg
Target Ras GTPase Inhibitors
Molecular Formula C30H30F2N6O3
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.