missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AMG 510

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
AMG 510 is a potent and selective covalent KRASG12C inhibitor (IC50 = 90 nM). Also a potent inhibitor of phospho-ERK (IC50 = 68 nM). In an in vivo model of non-small cell lung cancer, AMG 510 causes regression of tumors, an increase in T-cell recruitment and a pro-inflammatory tumor microenvironment. Synergises with MEK inhibitors and Carboplatin (Cat. No. 2626) and is orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | (1 R)-6-Fluoro-7-(2-fluoro-6-hydroxyphenyl)-1-[4-methyl-2-(1-methylethyl)-3-pyridinyl]-4-[(2 S)-2-methyl-4-(1-oxo-2-propen-1-yl)-1-piperazinyl]pyrido[2,3-d]pyrimidin-2(1 H)-one |
| CAS | 2252403-56-6 |
| Quantity | 50 mg |
| Target | Ras GTPase Inhibitors |
| Molecular Formula | C30H30F2N6O3 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction