missing translation for 'onlineSavingsMsg'
Learn More

Amido Black 10B, Fisher BioReagents

Product Code. 11433783
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433783 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11433783 Supplier Fisher BioReagents Supplier No. BP12410

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Amido Black 10B is a general stain for proteins and nucleic acids.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1064-48-8
Extinction Coefficient 450g(-1) cm(-1) min.
Molecular Formula C22H14N6Na2O9S2
Synonym Naphthol Blue Black
Identification Pass Test
InChI Key HKBVRFLHNUEVRO-UHFFFAOYSA-L
SMILES C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
Molecular Weight (g/mol) 616.487
PubChem CID 44134531
Lambda Max 616 to 620nm
Formula Weight 616.5
Loss on Drying 10% max.
Packaging Glass Bottle
Color Index No. 20470
Color Brown
pH 9.0
Quantity 10 g
Physical Form Powder
Chemical Name or Material Amido Black 10B
Show More Show Less
compliance-icons
Product Identifier
  • AMIDO BLACK 10B
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.