missing translation for 'onlineSavingsMsg'
Learn More

Amido Black 10B

Product Code. 11404320
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11404320 25 g 25g
11414320 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11404320 Supplier Thermo Scientific Alfa Aesar Supplier No. A11374.14
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used as a histological, protein, and nucleic acid dye used in electrophoresis. | CAS: 1064-48-8 | C22H14N6Na2O9S2 | 616.487 g/mol

Amido Black 10B is used as a histological, protein, and nucleic acid dye used in electrophoresis. Naphthol Blue Black is an important and commonly used industrial dye. This dye is also used in rapid staining of protein bands on nitrocellulose membranes such as Western blots. Naphthol Blue Black can be utilized as a nucleic acid stain in SDS-Page gels. Additionally this compound has been untilized in cell proliferation assays and histochemical studies. It is also used in criminal investigations to detect blood present with latent fingerprints. They are also used in paints, inks, plastics and leather.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1064-48-8
Molecular Formula C22H14N6Na2O9S2
Molecular Weight (g/mol) 616.487
MDL Number MFCD00004017
InChI Key HKBVRFLHNUEVRO-UHFFFAOYSA-L
Synonym C.I. 20470,Naphthalene Black 10B
PubChem CID 44134531
IUPAC Name disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Packaging Glass bottle
Sensitivity Hygroscopic
Beilstein 5374263
Solubility Information It is soluble in water (10mg/ml at 25°C),and ethanol.
Quantity 25 g
Physical Form Powder
Formula Weight 616.49
Chemical Name or Material Amido Black 10B
compliance-icons
Product Identifier
  • Amido Black 10B
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: C.I. Acid black 1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.