missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Amies Agar Gel with Charcoal Transport Swabs

Product Code. 10188242
Change view
Click to view available options
Quantity:
50/Pk., 500/Cs.
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10188242 50/Pk., 500/Cs. Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10188242 Supplier Thermo Scientific™ Supplier No. TS0002A
  • ONLINE EXCLUSIVE PRICE
    954.75 €
    897.48 € / Pack of 500
    Save 57.27 €
    5% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Simplify the transportation of throat, vaginal, wound and skin swabs with this Amies Agar Gel Transport Swabs.

Simplify the transportation of throat, vaginal, wound and skin swabs with Thermo Scientific™ Oxoid™ Amies Agar Gel with Charcoal Transport Swabs. The presence of charcoal in the medium neutralizes bacterial toxins and other inhibitory substances and prolongs the viability of pathogenic organisms. Amies Agar Gel with Charcoal Transport Swabs come with soft rayon tips, which are inert and non-toxic to microorganisms.

Ensure optimal sample collection and recovery using Amies Agar Gel with Charcoal Transport Swabs.

  • Contains media for the maintenance of bacterial sample during transport to the laboratory.
  • Each batch of product is tested for performance using a wide range of aerobic and anaerobic pathogens to ensure adequate recoveries.
  • Lot number/expiry date and tamper-evident tube ensure complete traceability and sterility.
  • “Venturi” Hour-Glass Tube Design prevents disintegration of gel column and eliminates undesirable air pockets, bubbles and breaks in gel which may be harmful to fastidious bacteria.
  • Maximum gel for anerobe preservations at 5mL provides sample viability and protection
  • Product does not contain glass ampules, meaning more safety as there is no need to break any glass and no seals or barriers to puncture or dry out swabs.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Product Type Transport Swab
Swab Configuration Single Applicator
Description Amies Agar Gel with Charcoal Transport Swabs
Quantity 50/Pk., 500/Cs.
Shaft Material Plastic
Storage Requirements 5°C to 25°C; do not freeze
Tip Material Rayon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.