missing translation for 'onlineSavingsMsg'
Learn More

Amifostine Thiol Dihydrochloride >90%, TRC

Product Code. 30009386
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009386 50 mg 50mg
30007468 100 mg 100mg
30006583 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009386 Supplier Toronto Research Chemicals Supplier No. TRCA57682050MG
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14653-77-1
Molecular Formula C5 H14 N2 S . 2 Cl H
Molecular Weight (g/mol) 207.16
Synonym 2-[(3-Aminopropyl)amino]ethanethiol Hydrochloride, WR 1065
IUPAC Name 2-(3-aminopropylamino)ethanethiol;dihydrochloride
SMILES Cl.Cl.NCCCNCCS

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C5H14N2S.2ClH/c6-2-1-3-7-4-5-8;;/h7-8H,1-6H2;2*1H
Formula Weight 206.0411
Chemical Name or Material Amifostine Thiol 2-[(3-aminopropyl)amino]ethanethiol, dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.