missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amifostine Trihydrate, TRC
Specifications
Specifications
| Chemical Name or Material | Amifostine trihydrate |
| CAS | 112901-68-5 |
| Molecular Formula | C5 H15 N2 O3 P S . 3 H2 O |
| Quantity | 100 mg |
| Synonym | 2-[(3-Aminopropyl)amino]ethanethiol dihydrogen phosphate (ester) trihydrate, Amifostine trihydrate, Amifostine |
| InChI Formula | InChI=1S/C5H15N2O3PS.3H2O/c6-2-1-3-7-4-5-12-11(8,9)10;;;/h7H,1-6H2,(H2,8,9,10);3*1H2 |
| SMILES | O.O.O.NCCCNCCSP(=O)(O)O |
| IUPAC Name | 2-(3-aminopropylamino)ethylsulfanylphosphonic acid;trihydrate |
| Molecular Weight (g/mol) | 268.27 |
| Formula Weight | 268.0858 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?