missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Amifostine trihydrate

Product Code. 16631852 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
16631852 250 mg 250mg
16621852 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 16631852

Brand: Thermo Scientific Chemicals 463102500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112901-68-5 | C5H21N2O6PS | 268.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112901-68-5
Molecular Formula C5H21N2O6PS
Molecular Weight (g/mol) 268.26
InChI Key TXQPXJKRNHJWAX-UHFFFAOYSA-N
IUPAC Name ({2-[(3-aminopropyl)amino]ethyl}sulfanyl)phosphonic acid trihydrate
SMILES O.O.O.NCCCNCCSP(O)(O)=O

Specifications

Quantity 250 mg
Formula Weight 268.27
Chemical Name or Material Amifostine trihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.