missing translation for 'onlineSavingsMsg'
Learn More

Amikacin disulfate salt, Thermo Scientific Chemicals

Product Code. 15589343 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15589343 5 g 5g
15599343 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15589343 Supplier Thermo Scientific Acros Supplier No. 455190050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39831-55-5
Molecular Formula C22H47N5O21S2
Molecular Weight (g/mol) 781.75
MDL Number MFCD00167475,MFCD00167475
InChI Key FXKSEJFHKVNEFI-GCZBSULCSA-N
PubChem CID 134129615
IUPAC Name (2S)-4-amino-N-[(1R,2S,3S,4S,5S)-5-amino-2-[(2S,3S,4S,5S,6S)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4-[(2R,3S,4S,5S,6S)-6-(aminomethyl)-3,4,5-trihydroxyoxan-2-yl]oxy-3-hydroxycyclohexyl]-2-hydroxybutanamide;sulfuric acid
SMILES OS(O)(=O)=O.OS(O)(=O)=O.NCC[C@H](O)C(=O)N[C@@H]1C[C@H](N)[C@@H](O[C@H]2O[C@H](CN)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O[C@H]1O[C@H](CO)[C@@H](O)[C@H](N)[C@H]1O

Specifications

Melting Point 220.0°C to 230.0°C
pH 2 to 4 (1% aq. soln.)
Loss on Drying 13% max. (110°C, 3 h, 5 mm Hg)
Infrared Spectrum Authentic
Quantity 5 g
Solubility Information Solubility in water: ca 50g/L.
Formula Weight 781.76
Chemical Name or Material Amikacin disulfate salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.