missing translation for 'onlineSavingsMsg'
Learn More

Amikacin disulfate, Thermo Scientific™

An aminoglycoside antibiotic derived from Kanamycin A

155.00 € - 445.00 €

Chemical Identifiers

CAS 39831-55-5
Molecular Formula C22H47N5O21S2
Molecular Weight (g/mol) 781.75
MDL Number MFCD00167475,MFCD00167475
InChI Key FXKSEJFHKVNEFI-GCZBSULCSA-N
PubChem CID 134129615
IUPAC Name (2S)-4-amino-N-[(1R,2S,3S,4R,5S)-5-amino-2-{[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4-{[(2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxyoxan-2-yl]oxy}-3-hydroxycyclohexyl]-2-hydroxybutanamide; bis(sulfuric acid)
SMILES OS(O)(=O)=O.OS(O)(=O)=O.NCC[C@H](O)C(=O)N[C@@H]1C[C@H](N)[C@@H](O[C@H]2O[C@H](CN)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O[C@H]1O[C@H](CO)[C@@H](O)[C@H](N)[C@H]1O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15416019
SDS View Documents
Thermo Scientific Alfa Aesar
J63862.14
25g
155.00 €
25g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
15426019
SDS View Documents
Thermo Scientific Alfa Aesar
J63862.22
445.00 €
100g
View alternative products
Save Up to 
»
Estimated Shipment: 10-09-2026
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Amikacin disulfate is used as an aminoglycoside antibiotic derived from Kanamycin A, used for treating infections with multidrug resistant Gram negative bacteria such as Pseudomonas aeruginosa, Acinetobacter, and Enterobacter. It is commonly used in the treatment of drug-resistant mycobacteria. It is used to study organism-directed delivery of antibiotics as well as drug resistance.

Solubility
Soluble in water at 50mg/ml with warming

Notes
Potency: 674-786 micrograms per mg
Specifications

Chemical Identifiers

39831-55-5
781.75
FXKSEJFHKVNEFI-GCZBSULCSA-N
(2S)-4-amino-N-[(1R,2S,3S,4R,5S)-5-amino-2-{[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4-{[(2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxyoxan-2-yl]oxy}-3-hydroxycyclohexyl]-2-hydroxybutanamide; bis(sulfuric acid)
C22H47N5O21S2
MFCD00167475,MFCD00167475
134129615
OS(O)(=O)=O.OS(O)(=O)=O.NCC[C@H](O)C(=O)N[C@@H]1C[C@H](N)[C@@H](O[C@H]2O[C@H](CN)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O[C@H]1O[C@H](CO)[C@@H](O)[C@H](N)[C@H]1O

Specifications

39831-55-5
C22H47N5O21S2
6172633
Soluble in water at 50mg/ml with warming
OS(O)(=O)=O.OS(O)(=O)=O.NCC[C@H](O)C(=O)N[C@@H]1C[C@H](N)[C@@H](O[C@H]2O[C@H](CN)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O[C@H]1O[C@H](CO)[C@@H](O)[C@H](N)[C@H]1O
781.75
781.77
White
MFCD00167475,MFCD00167475
14,405
FXKSEJFHKVNEFI-GCZBSULCSA-N
(2S)-4-amino-N-[(1R,2S,3S,4R,5S)-5-amino-2-{[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4-{[(2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxyoxan-2-yl]oxy}-3-hydroxycyclohexyl]-2-hydroxybutanamide; bis(sulfuric acid)
134129615
Amikacin disulfate
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 254-648-6

missing translation for 'rtecsNumber' : WK1961200

missing translation for 'tsca' : No

Recommended Storage : Keep cold

Product Identifier
  • Amikacin disulfate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contain: Amikacin sulfate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.