missing translation for 'onlineSavingsMsg'
Learn More

Amikacin disulfate

Product Code. 15426019
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15426019 100 g 100g
15416019 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15426019 Supplier Thermo Scientific Alfa Aesar Supplier No. J63862.22
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An aminoglycoside antibiotic derived from Kanamycin A | CAS: 39831-55-5 | C22H47N5O21S2 | 781.75 g/mol

Amikacin disulfate is used as an aminoglycoside antibiotic derived from Kanamycin A, used for treating infections with multidrug resistant Gram negative bacteria such as Pseudomonas aeruginosa, Acinetobacter, and Enterobacter. It is commonly used in the treatment of drug-resistant mycobacteria. It is used to study organism-directed delivery of antibiotics as well as drug resistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amikacin disulfate is used as an aminoglycoside antibiotic derived from Kanamycin A, used for treating infections with multidrug resistant Gram negative bacteria such as Pseudomonas aeruginosa, Acinetobacter, and Enterobacter. It is commonly used in the treatment of drug-resistant mycobacteria. It is used to study organism-directed delivery of antibiotics as well as drug resistance.

Solubility
Soluble in water at 50mg/ml with warming

Notes
Potency: 674-786 micrograms per mg
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39831-55-5
Molecular Formula C22H47N5O21S2
Molecular Weight (g/mol) 781.75
MDL Number MFCD00167475,MFCD00167475
InChI Key FXKSEJFHKVNEFI-GCZBSULCSA-N
PubChem CID 134129615
IUPAC Name (2S)-4-amino-N-[(1R,2S,3S,4R,5S)-5-amino-2-{[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4-{[(2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxyoxan-2-yl]oxy}-3-hydroxycyclohexyl]-2-hydroxybutanamide; bis(sulfuric acid)
SMILES OS(O)(=O)=O.OS(O)(=O)=O.NCC[C@H](O)C(=O)N[C@@H]1C[C@H](N)[C@@H](O[C@H]2O[C@H](CN)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O[C@H]1O[C@H](CO)[C@@H](O)[C@H](N)[C@H]1O

Specifications

Type Amikacin Disulfate
Physical Form Powder
Melting Point 220°C to 230°C
Color White
Quantity 100 g
Beilstein 6172633
Merck Index 14,405
Solubility Information Soluble in water at 50mg/ml with warming
Formula Weight 781.77
Chemical Name or Material Amikacin disulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.