missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Amine-PEG3-Biotin

Product Code. 16460747
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16460747 50 mg 50mg
16457157 5 mg 5mg
16425326 100 mg 100mg
16414438 10 mg 10mg
16413086 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16460747 Supplier MedChemExpress Supplier No. HY111377
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Amine-PEG3-Biotin is a signal amplification label containing a biotin group and a terminal primary amine group.

Specifications

Quantity 50 mg
Molecular Weight (g/mol) 418.55 g/mol
Solubility DMSO : 250 mg/mL (597.30 mM; Need ultrasonic) ∣ H2O : 100 mg/mL (238.92 mM; Need ultrasonic)
Content And Storage 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Shipping Condition Room temperature
CAS 359860-27-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.