missing translation for 'onlineSavingsMsg'
Learn More

ε-Amino-N-Caproic Acid, MP Biomedicals™

Product Code. 11465965 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 kg
Unit Size:
5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465965 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11465965 Supplier MP Biomedicals Supplier No. 0210164594
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibits plasmin and carboxypeptidase B. ε-Amino-N-Caproic Acid, MP Biomedicals is a lysine analog that promotes rapid dissociation of plasmin, thereby inhibiting the activation of plasminogen and subsequent fibrinolysis. It is has been utilized i

EACA is reported to inhibit chymotrypsin, Factor VIIa, lysine carbo­xy­peptidase, plasmin, and plasminogen activator. Reported to inhibit plasminogen binding to activated platelets. An early report indicated that it inhibits the activation of the first component of the complement system. Binds and inactivates Carboxypeptidase B.

Specifications

CAS 60-32-2
Melting Point 205°C to 215°C
Density 1.48g/mL (lit.)
Molecular Formula C6H13NO2
Quantity 5 kg
Synonym 6-aminocaproic acid, aminocaproic acid, amicar, epsikapron, capramol, caprocid, epsamon, eaca, acepramin, caprolisin
Solubility Information Soluble in water (50mg/mL); sparingly soluble in alcohol and methanol; practically insoluble in ether; insoluble in ethanol.
InChI Key SLXKOJJOQWFEFD-UHFFFAOYSA-N
SMILES C(CCC(=O)O)CCN
IUPAC Name 6-aminohexanoic acid
Molecular Weight (g/mol) 131.175
PubChem CID 564
ChEBI CHEBI:16586
Formula Weight 131.2
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.