missing translation for 'onlineSavingsMsg'
Learn More

Aminoguanidine hydrogen carbonate, 98+%

Product Code. 11490578
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490578 2500 g 2500g
11470578 100 g 100g
11480578 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490578 Supplier Thermo Scientific Alfa Aesar Supplier No. B24696.0E
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2582-30-1 | C2H8N4O3 | 136.111 g/mol

Aminoguanidine bicarbonate is an inhibitor of NOS (nitric oxide synthase). Aminoguanidine bicarbonate is used to study the effect of addition of polyamines to rat embryo cell cultures infected with adenovirus type 5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aminoguanidine bicarbonate is an inhibitor of NOS (nitric oxide synthase). Aminoguanidine bicarbonate is used to study the effect of addition of polyamines to rat embryo cell cultures infected with adenovirus type 5.

Solubility
Soluble in water (3.3 mg/ml at 30°C), and water (2.7 mg/ml at 20°C).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2582-30-1
Molecular Formula C2H8N4O3
Molecular Weight (g/mol) 136.111
MDL Number MFCD00012949
InChI Key OTXHZHQQWQTQMW-UHFFFAOYSA-N
Synonym aminoguanidine bicarbonate, aminoguanidine hydrogen carbonate, aminoguanidinium bicarbonate, aminoguanidine hydrocarbonate, aminoguanidine carbonate 1:1, aminoguanidium hydrogen carbonate, n1-aminoguanidine carbonate 1:1, aminoguanidine hydrogencarbonate, aminoguanidinebicarbonate, hydrazinecarboximidamide carbonate
PubChem CID 164944
IUPAC Name 2-aminoguanidine;carbonic acid
SMILES C(=NN)(N)N.C(=O)(O)O

Especificaciones

Melting Point ∼170°C (decomposition)
Density 1.6
Quantity 2500 g
UN Number UN3077
Beilstein 3569869
Solubility Information Soluble in water (3.3mg/ml at 30°C),and water (2.7mg/ml at 20°C).
Formula Weight 136.11
Percent Purity ≥98%
Chemical Name or Material Aminoguanidine hydrogen carbonate

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.