missing translation for 'onlineSavingsMsg'
Learn More

Aminoguanidine nitrate, 98%, cont. ca 10% water

Product Code. 11332537
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332537 25 g 25g
11342537 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332537 Supplier Thermo Scientific Alfa Aesar Supplier No. L11632.14
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 10308-82-4 | CH7N5O3 | 137.10 g/mol

Aminoguanidine nitrate is used as a pharmaceutical raw material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aminoguanidine nitrate is used as a pharmaceutical raw material.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with reducing agents, strong oxidizing agents, organic materials and metal powders.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10308-82-4
Molecular Formula CH7N5O3
Molecular Weight (g/mol) 137.10
MDL Number MFCD00013174
InChI Key PMGFHEJUUBDCLU-UHFFFAOYSA-N
Synonym aminoguanidine nitrate, aminoguanidine, nitrate, hydrazinecarboximidamide, mononitrate, monoaminoguanidium nitrate, aminoguanidine, mononitrate, hydrazinecarboximidamide nitrate, hydrazinecarboximidamide, nitrate 1:1, 1-aminoguanidine; nitric acid, aminoguanidine; nitric acid
PubChem CID 165859
IUPAC Name 2-aminoguanidine;nitric acid
SMILES O[N+]([O-])=O.NN=C(N)N

Specifications

Melting Point 138°C to 142°C
Quantity 25 g
UN Number UN1479
Beilstein 3916056
Solubility Information Soluble in water.
Formula Weight 137.1
Percent Purity 98%
Assay cont. ≈10% water
Chemical Name or Material Aminoguanidine nitrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.