missing translation for 'onlineSavingsMsg'
Learn More

Aminoguanidine nitrate, 99%

Product Code. 10560584 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10560584 25 g Plastic bottle 25g
10254510 5 g Glass bottle 5g
10142800 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10560584 Supplier Thermo Scientific Acros Supplier No. 103760250
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10308-82-4 | CH7N5O3 | 137.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10308-82-4
Molecular Formula CH7N5O3
Molecular Weight (g/mol) 137.10
MDL Number MFCD00013174
InChI Key PMGFHEJUUBDCLU-UHFFFAOYSA-N
Synonym aminoguanidine nitrate, aminoguanidine, nitrate, hydrazinecarboximidamide, mononitrate, monoaminoguanidium nitrate, aminoguanidine, mononitrate, hydrazinecarboximidamide nitrate, hydrazinecarboximidamide, nitrate 1:1, 1-aminoguanidine; nitric acid, aminoguanidine; nitric acid
PubChem CID 165859
IUPAC Name 2-aminoguanidine;nitric acid
SMILES O[N+]([O-])=O.NN=C(N)N

Specifications

Melting Point 145°C to 147°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula H2NNHC(=NH)NH2·HNO3
Quantity 25 g
Beilstein 03,117
Formula Weight 137.11
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Aminoguanidine nitrate, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Aminoguanidine nitrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.