missing translation for 'onlineSavingsMsg'
Learn More

Aminoguanidine Nitrate, TRC

Product Code. 30321890
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30321890 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30321890 Supplier Toronto Research Chemicals Supplier No. TRCA60972010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Aminoguanidinium nitrate
CAS 10308-82-4
Molecular Formula C H7 N4 . N O3
Quantity 10 g
Synonym Hydrazinecarboximidamide Mononitrate, Aminoguanidine Mononitrate, Aminoguanidine Nitrate, Aminoguanidinium Nitrate, N-Aminoguanidine Nitrate, NSC 206198
InChI Formula InChI=1S/CH6N4.NO3/c2-1(3)5-4;2-1(3)4/h4H2,(H4,2,3,5);/q;-1/p+1
SMILES NNC(=[NH2+])N.[O-][N+](=O)[O-]
IUPAC Name [amino(hydrazinyl)methylidene]azanium;nitrate
Molecular Weight (g/mol) 137.1
Formula Weight 137.0549
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.