missing translation for 'onlineSavingsMsg'
Learn More

Aminomethanephosphonic Acid, TRC

Product Code. 17994005
Change view
Click to view available options
Quantity:
100 mg
250 mg
5 g
Unit Size:
100mg
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17994005 250 mg 250mg
17962835 5 g 5g
17980505 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17994005 Supplier Toronto Research Chemicals Supplier No. TRCA617340250MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1066-51-9
Molecular Formula C H6 N O3 P
Molecular Weight (g/mol) 111.04
Synonym Phosphonic acid, (aminomethyl)- (6CI,7CI,8CI,9CI), P-(Aminomethyl)phosphonic acid, (Aminomethyl)phosphonic acid, AMPA, AMPA (surfactant), Aminomethanephosphonic acid, Aminomethylenephosphonic acid, NSC 30076, Nalco 3919, Phosphaglycine
IUPAC Name aminomethylphosphonic acid
SMILES NCP(=O)(O)O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/CH6NO3P/c2-1-6(3,4)5/h1-2H2,(H2,3,4,5)
Formula Weight 111.0085
Percent Purity >95%
Chemical Name or Material Aminomethyl phosphonic acid (AMPA)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.