missing translation for 'onlineSavingsMsg'
Learn More

(Aminomethyl)cyclopropane hydrochloride, 97%, Thermo Scientific™

Product Code. 11985191 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11985191 1 g 1g
11905201 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11985191 Supplier Thermo Scientific Acros Supplier No. 104010010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7252-53-1
Molecular Formula C4H9ClN
Molecular Weight (g/mol) 106.57
MDL Number MFCD00012544
InChI Key MQCZBGGAZQXTLH-UHFFFAOYSA-N
Synonym cyclopropanemethylamine hydrochloride, cyclopropylmethanamine hydrochloride, cyclopropylmethyl ammonium chloride, cyclopropylmethylamine hydrochloride, aminomethylcyclopropane hydrochloride, 1-cyclopropylmethanamine hydrochloride, aminomethyl cyclopropane hydrochloride, cyclopropylmethanaminium chloride, cyclopropylmethylamin hydrochloride, cyclopropylmethanamine-hydrochloride
PubChem CID 81671
SMILES [Cl].NCC1CC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 205.0°C to 209.0°C
Color White
Elemental Analysis 42.87 to 46.44% C, 12.50 to 13.54% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula C3H5CH2NH2·HCl
Quantity 1 g
Beilstein 12,4
IUPAC Name 1-cyclopropylmethanamine hydrochloridyl
Formula Weight 107.58
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (Aminomethyl)cyclopropane hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • (Aminomethyl)cyclopropane hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.