missing translation for 'onlineSavingsMsg'
Learn More

(Aminomethyl)phosphonic acid, 99%, Thermo Scientific Chemicals

Product Code. 10380684 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10380684 250 mg Glass bottle 250mg
10732681 1 g Glass bottle 1g
10341434 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10380684 Supplier Thermo Scientific Acros Supplier No. 291642500
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1066-51-9
Molecular Formula CH6NO3P
Molecular Weight (g/mol) 111.04
MDL Number MFCD00008105
InChI Key MGRVRXRGTBOSHW-UHFFFAOYSA-N
Synonym aminomethyl phosphonic acid, aminomethanephosphonic acid, amep, 1-aminomethylphosphonic acid, 1-aminomethylphosphonate, amino methane phosphoric acid, caswell no. 037c, 1-aminomethyl phosphonic acid, phosphonic acid, aminomethyl
PubChem CID 14017
ChEBI CHEBI:28812
IUPAC Name aminomethylphosphonic acid
SMILES C(N)P(=O)(O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula H2NCH2P(O)(OH)2
Quantity 250 mg
Beilstein 01, 2596
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohols
Formula Weight 111.04
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material (Aminomethyl)phosphonic acid
Show More Show Less
compliance-icons
Product Identifier
  • (Aminomethyl)phosphonic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.