missing translation for 'onlineSavingsMsg'
Learn More

Aminomethyl phosphonic acid (AMPA) 13C 15N D2 100 μg/mL in Water, Dr. Ehrenstorfer

Product Code. 30319894
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30319894 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30319894 Supplier Dr. Ehrenstorfer Supplier No. DREA10205120WA100
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aminomethylphosphonic acid 13C, 15N, methylene-d2
CAS 2483829-28-1
Solvent or Matrix Water
Molecular Formula 13C 2H2 H4 15N O3 P
Quantity 1 mL
Synonym AMPA 13C, 15N, d2
InChI Formula InChI=1S/CH6NO3P/c2-1-6(3,4)5/h1-2H2,(H2,3,4,5)/i1+1D2,2+1
SMILES [2H][13C]([2H])([15NH2])P(=O)(O)O
IUPAC Name [azanyl(dideuterio)(1^{13}C)methyl]phosphonic acid
Molecular Weight (g/mol) 115.04
Formula Weight 115.0215 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.