missing translation for 'onlineSavingsMsg'
Learn More

Aminopyralid, TRC

Product Code. 30311136
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30311136 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30311136 Supplier Toronto Research Chemicals Supplier No. TRCA628820250MG
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aminopyralid
CAS 150114-71-9
Purity Grade Notes HPLC
Molecular Formula C6 H4 Cl2 N2 O2
Quantity 250 mg
Synonym 4-Amino-3,6-dichloro-2-pyridinecarboxylic acid, 4-Amino-3,6-dichloropicolinic acid, 4-Amino-3,6-dichloropyridine-2-carboxylic acid, Aminopyralid, Milestone, Milestone (aminopyralid herbicide)
InChI Formula InChI=1S/C6H4Cl2N2O2/c7-3-1-2(9)4(8)5(10-3)6(11)12/h1H,(H2,9,10)(H,11,12)
SMILES Nc1cc(Cl)nc(C(=O)O)c1Cl
IUPAC Name 4-amino-3,6-dichloropyridine-2-carboxylic acid
Molecular Weight (g/mol) 207.01
Formula Weight 205.965
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.