missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Amisulpride
Description
Amisulpride is a potent, selective dopamine D2 and D3 receptor antagonist. Ki values are 2.8 and 3.2 nM respectively for human D2 and D3 and > 1000 nM for human D1, D4 and D5 receptors. Shows selectivity for presynaptic dopamine autoreceptors at low doses and blocks postsynaptic D2/D3 receptors at higher doses. Preferentially interacts with limbic D2-like receptors in vivo. Atypical antipsychotic/antischizophrenic agent with limited extrapyrimidal side effects and a profile distinct from that of haloperidol and remoxipride.
Specifications
Specifications
| Purity | >98% |
| Gene Alias | DAN 2163 |
| Quantity | 50 mg |
| Product Type | Amisulpride |
| Molecular Weight (g/mol) | 369.48 |
| Product Line | Tocris |
| Inhibitors | Amisulpride |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?