missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amlexanox, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | Amlexanox |
| CAS | 68302-57-8 |
| Molecular Formula | C16 H14 N2 O4 |
| Quantity | 500 mg |
| Synonym | Amlexanox - Bio-X TM |
| InChI Formula | InChI=1S/C16H14N2O4/c1-7(2)8-3-4-12-9(5-8)13(19)10-6-11(16(20)21)14(17)18-15(10)22-12/h3-7H,1-2H3,(H2,17,18)(H,20,21) |
| SMILES | CC(C)c1ccc2Oc3nc(N)c(cc3C(=O)c2c1)C(=O)O |
| IUPAC Name | 2-amino-5-oxo-7-propan-2-ylchromeno[2,3-b]pyridine-3-carboxylic acid |
| Molecular Weight (g/mol) | 298.29 |
| Formula Weight | 298.0954 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction