missing translation for 'onlineSavingsMsg'
Learn More

Amlodipine Besylate, TRC

Product Code. 17971935
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17971935 5 g 5g
17942715 100 mg 100mg
17952365 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17971935 Supplier Toronto Research Chemicals Supplier No. TRCA6335005G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111470-99-6
Molecular Formula C20 H25 Cl N2 O5 . C6 H6 O3 S
Molecular Weight (g/mol) 567.05
Synonym 3,5-Pyridinedicarboxylic acid, 2-[(2-aminoethoxy)methyl]-4-(2-chlorophenyl)-1,4-dihydro-6-methyl-, 3-ethyl 5-methyl ester, monobenzenesulfonate (9CI), Benzenesulfonic acid, compd. with 3-ethyl 5-methyl 2-[(2-aminoethoxy)methyl]-4-(2-chlorophenyl)-1,4-dihydro-6-methyl-3,5-pyridinedicarboxylate (1:1), (±)-3-Ethyl 5-methyl 2-[(2-aminoethoxy)methyl]-4-(2-chlorophenyl)-1,4-dihydro-6-methyl-3,5-pyridinedicarboxylate monobenzenesulfonate, (±)-Amlodipine besylate, AM Arya 5, Alkapress, Alopres, Amcard, Amdepin, Amdipin, Amilo, Amlodin, Amlodin OD, Amlodipine benzenesulfonate, Amlodipine benzenesulfonate salt, Amlodipine besylate, Amlodis, Amlogard, Amlopers 5, Amlopin, Amlor, Amlosyn, Amlovas, Antacal, Calchek, Cardilopin, Istin, Lowrac, Monopina, Myodura, Normodipine, Norvas, Norvasc, Norvask, Regcor, Tensivask, UK 48340-26, Vazkor
IUPAC Name benzenesulfonic acid;3-O-ethyl 5-O-methyl 2-(2-aminoethoxymethyl)-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=C(COCCN)NC(=C(C1c2ccccc2Cl)C(=O)OC)C.OS(=O)(=O)c3ccccc3

Specifications

Quantity 5 g
InChI Formula InChI=1S/C20H25ClN2O5.C6H6O3S/c1-4-28-20(25)18-15(11-27-10-9-22)23-12(2)16(19(24)26-3)17(18)13-7-5-6-8-14(13)21;7-10(8,9)6-4-2-1-3-5-6/h5-8,17,23H,4,9-11,22H2,1-3H3;1-5H,(H,7,8,9)
Formula Weight 566.149
Percent Purity >95%
Chemical Name or Material Amlodipine besilate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.