missing translation for 'onlineSavingsMsg'
Learn More

WTW™ Ammonia and Ammonium Test Kits

Product Code. p-8001768
Click to view available options
Type:
Ammonium (NH4) Reagent Test Kit
Unit Size:
Pack of 25
This item is not returnable. View return policy

Product Code. 11794745

Brand: WTW™ 250424

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits

TRUSTED_SUSTAINABILITY

Specifications

Analytes Salyticate
For Use With (Application) For waste water, drinking water, sea water
For Use With (Equipment) PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, spectroFlex photometers
Includes Certificates
No. of Tests 950
Range 4 to 80 mg/L NH4-N, 5.2 to 103 mg/L NH4+
Sufficient For 100 Tests
Test Method Photometric
Type Ammonium (NH4) Reagent Test Kit
compliance-icons
Product Identifier
  • Ammonium Cell Test Method: photometric 4.0 - 80.0 mg/l NH
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contains : Sodium nitroprusside, troclosene sodium,dihydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.