missing translation for 'onlineSavingsMsg'
Learn More

Ammonia Solution, 0.88 S.G., 1:2 Dilution, Fisher Chemical

Product Code. 12696727 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12696727 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12696727 Supplier Fisher Chemical Supplier No. J/9015/08
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

pH 12
Color Colorless
Boiling Point 38°C
CAS 1336-21-6
Melting Point -57°C
Molecular Formula H5NO
MDL Number MFCD00066650
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Quantity 500 mL
SMILES N.O
IUPAC Name azanium;hydroxide
Molecular Weight (g/mol) 35.05
PubChem CID 14923
ChEBI CHEBI:18219
Packaging HDPE plastic bottle
Vapor Density 0.59
Vapor Pressure 500 hPa at 20°C
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ammonia solution, 0.88 S.G., 1:2 dilution in water
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contain: Ammonium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.