missing translation for 'onlineSavingsMsg'
Learn More

Ammonia Solution, 2M in Ethanol, Honeywell Fluka™

Product Code. 15635360 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15635360 100 mL Glass Bottle 100mL
15645360 100 mL Glass Bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15635360 Supplier Honeywell-Fluka Supplier No. 392685100ML
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

2.0 M in ethanol

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-41-7
Molecular Formula H3N
Molecular Weight (g/mol) 17.03
MDL Number MFCD00011418
InChI Key QGZKDVFQNNGYKY-UHFFFAOYSA-N
Synonym ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-fol
PubChem CID 222
ChEBI CHEBI:16134
SMILES N

Specifications

Boiling Point 60°C
Linear Formula NH3
Concentration 2M in ethanol
UN Number UN1170
Density 0.785g/mL at 25°C
Quantity 100 mL
IUPAC Name ammonia
Flash Point 3°C (37.4°F)
Formula Weight 17.03g/mol
Packaging Glass Bottle
Physical Form Liquid
Chemical Name or Material Ammonia solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.