missing translation for 'onlineSavingsMsg'
Learn More

Ammonia Solution, 2M in Isopropanol, Honeywell Fluka™

Product Code. 15665360 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15665360 800 mL Glass Bottle 800mL
15655360 100 mL Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15665360 Supplier Honeywell-Fluka Supplier No. 392693800ML
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

2.0 M in isopropanol

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-41-7
Molecular Formula H3N
Molecular Weight (g/mol) 17.03
MDL Number MFCD00011418
InChI Key QGZKDVFQNNGYKY-UHFFFAOYSA-N
Synonym ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-fol
PubChem CID 222
ChEBI CHEBI:16134
SMILES N

Especificaciones

Boiling Point 60°C
Linear Formula NH3
Concentration 2.0M in isopropanol
UN Number UN1219
Density 0.773g/mL at 25°C
Quantity 800 mL
IUPAC Name azane
Flash Point 4°C (39.2°F)
Formula Weight 17.03g/mol
Packaging Glass Bottle
Vapor Pressure 6.34psi (20°C), 20.22psi (55°C)
Physical Form Liquid
Chemical Name or Material Ammonia solution
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.