missing translation for 'onlineSavingsMsg'
Learn More

Ammonia solution, 2.0 M in methanol, Honeywell Fluka™

Product Code. 15694110 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 L
4 x 100 mL
Packaging:
Glass Bottle
Unit Size:
100mL
2 L
800mL
Pack of 4
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15694110 4 x 100 mL Glass Bottle Pack of 4
15674110 100 mL Glass Bottle 100mL
15604120 800 mL Glass Bottle 800mL
15684110 2 L Glass Bottle 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15694110 Supplier Honeywell-Fluka Supplier No. 3414284X100ML
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

2.0 M in methanol

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-41-7 , 67-56-1
Molecular Formula H3N
Molecular Weight (g/mol) 17.03
MDL Number MFCD00011418
InChI Key QGZKDVFQNNGYKY-UHFFFAOYSA-N
Synonym ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-fol
PubChem CID 222
ChEBI CHEBI:16134
IUPAC Name ammonia
SMILES N

Specifications

CAS Max % 96.5940%, 3.4060%
Quantity 4 x 100 mL
Linear Formula NH3
UN Number UN1992
Flash Point 6°C
Packaging Glass Bottle
Density 0.787 g/mL at 25 °C
Chemical Name or Material Ammonia solution, 2.0 M in methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.