missing translation for 'onlineSavingsMsg'
Learn More

Ammonia Solution, 35%, Certified AR for Analysis, d=0.88, Fisher Chemical™

Product Code. 10111660 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
HDPE plastic bottle
Plastic Coated Glass bottle
Plastic container
Unit Size:
1 L
2.5 L
25 L
5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10111660 2.5 L Plastic Coated Glass bottle 2.5 L
10305220 1 L Plastic Coated Glass bottle 1 L
10049602 5 L HDPE plastic bottle 5 L
10659263 25 L Plastic container 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10111660 Supplier Fisher Chemical Supplier No. A/3280/PB17
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

H5NO, CAS Number-1336-21-6, aquammonia, ammonium hydroxide solution, ammonium hydroxide nh4 oh, ammonia water, aqua ammonia, ammonia, monohydrate, ammonium hydroxide, ammonia aqueous, household ammonia, ammonia, aqua, 2.5L, CHEBI:18219, Colorless, 35.05g/mol, VHUUQVKOLVNVRT-UHFFFAOYSA-N

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
SMILES N.O

Spezifikation

pH 12
Color Colorless
Quantity 2.5 L
IUPAC Name amine hydrate
Formula Weight 35.05g/mol
Packaging Plastic Coated Glass bottle
Physical Form Liquid
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Ammonia solution 35%
Signalwort
  • Danger
Gefahrenkategorie
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Gefahrenhinweis
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Ergänzende Informationen
  • MIXTURE LIST-Contain: Ammonium hydroxide,Ammonia
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.