missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Ammonium Acetate, Ultra Pure Grade

Product Code. 17138548 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17138548 500 g 500g
11456241 1 kg 1kg
17128548 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17138548 Supplier MP Biomedicals™ Supplier No. 0219384890
  • ONLINE EXCLUSIVE PRICE
    94.10 €
    89.25 € / 500g
    Save 4.85 €
    5% Off
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ammonium Acetate, Ultra Pure Grade is used in the purification and precipitation of DNA and for protein crystallization.

  • Ammonium acetate, ultra pure, is for applications which require tight control of elemental content.
  • Ammonium acetate is a widely used reagent in molecular biology and chromatography.
  • Its applications include the purification and precipitation of DNA and protein crystallization.
  • Ammonium acetate is commonly used in HPLC and MS analysis of various compounds, such as oligosaccharides, proteins, and peptides.
  • Ammonium acetate is also used for the nonaqueous capillary electrophoresis-mass spectrometry (NACE-MS) of lipophilic peptides and therapeutic drugs.

Specifications

Assay ≥97%
Chemical Name or Material Ammonium Acetate
CAS 631-61-8
Content And Storage Room Temperature
Density 1.17 at 68° F
Formula Weight 77.1
Melting Point 237.2° F
Molecular Formula C2H7NO2
Purity Ultrapure
Quantity 500 g
Synonym unii-rre756s6q2
Form Crystalline Powder
Show More Show Less

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.