missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Acetate-d7, CDN

Product Code. 30313712
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
missing translation for 'unitSize'
5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30313712

missing translation for 'mfr': CDN CDND59695G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material Ammonium Acetate-d7
CAS 194787-05-8
Molecular Formula C2 D3 O2 . D4 N
Quantity 5 g
Synonym Acetic-d3 acid-d, ammonium-d4 salt (9 CI), Ammonium-d4 acetate-d3, Ammonium acetate-d7, Ammonium acetate D7
InChI Formula InChI=1 S/C2H4O2.H3N/c1-2(3)4;/h1H3,(H,3,4);1H3/i1D3;/hD4
SMILES [2 H]C([2 H])([2 H])C(=O)[O-].[2 H][N+]([2 H])([2 H])[2 H]
IUPAC Name tetradeuterioazanium;2,2,2-trideuterioacetate
Molecular Weight (g/mol) 84.126
Formula Weight 84.092 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.