missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Acetate, For HPLC, Solstice

Product Code. 15681570
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15681570 50 g 50g
15671570 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15681570 Supplier Honeywell-Fluka Supplier No. 1783650G
Chemical Monthly Promotion
Use promo code 28093
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

for HPLC,≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.083
MDL Number MFCD00013066
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]

Specifications

Quantity 50 g
Linear Formula CH3CO2NH4
Beilstein 4186741
Solubility Information Soluble in water 0.1g/mL at 25°C
Formula Weight 77.08g/mol
Percent Purity ≥99%
Chemical Name or Material Ammonium Acetate
Product Identifier
  • Ammonium acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.