missing translation for 'onlineSavingsMsg'
Learn More

Ammonium acetate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥98%, Solstice

Product Code. 15623790
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
6 x 500 g
10 kg
20 kg
6 x 1 kg
6 x 2.5 kg
Unit Size:
100g
10kg
1kg
2.5kg
20kg
500g
Pack of 6
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15623790 6 x 1 kg Pack of 6
15663780 100 g 100g
15613790 500 g 500g
15683780 1 kg 1kg
15693780 2.5 kg 2.5kg
15643790 6 x 500 g Pack of 6
15673780 10 kg 10kg
15633790 6 x 2.5 kg Pack of 6
15603790 20 kg 20kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 15623790 Supplier Honeywell-Fluka Supplier No. 323016X1KG
  • ONLINE EXCLUSIVE PRICE
    740.00 €
    242.27 € / Pack of 6
    Save 497.73 €
    67% Off
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥98%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.083
MDL Number MFCD00013066
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]

Specifications

Melting Point 110°C to 112°C (decomposition)(literature)
CAS Max % ≤100.0000%
Physical Form Solid
Quantity 6 x 1 kg
Linear Formula CH3CO2NH4
Beilstein 4186741
Formula Weight 77.08
Percent Purity >98%
Grade ACS Reagent/Reagent Ph. Eur.
Packaging Poly Bottle
Density 1.170 g/cm3
Chemical Name or Material Ammonium acetate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.