missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Acetate Solution 0.25M, Certified, HPLC for Gradient Analysis, Fisher Chemical

Product Code. 10649633 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Plastic Coated Glass bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10649633 500 mL Plastic Coated Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10649633 Supplier Fisher Chemical Supplier No. J/0200/PB08
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 631-61-8
Color Colorless
Physical Form Liquid
Quantity 500 mL
Molecular Formula C2H7NO2
MDL Number 13066
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
SMILES CC(=O)[O-].[NH4+]
IUPAC Name azanium;acetate
Molecular Weight (g/mol) 77.083
PubChem CID 517165
ChEBI CHEBI:62947
Packaging Plastic Coated Glass bottle
Show More Show Less
Product Identifier
  • Ammonium acetate 0.25M for for HPLC method development
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.