missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Acetate, TraceSELECT™, Honeywell™

Product Code. 15656940 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 15656940

Brand: Honeywell-Fluka 73432100G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 631-61-8
Quantity 100 g
Molecular Formula C2H7NO2
Linear Formula CH3CO2NH4
MDL Number MFCD00013066
UN Number UN3077
Beilstein 4186741
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
SMILES CC(=O)[O-].[NH4+]
IUPAC Name azanium;acetate
Molecular Weight (g/mol) 77.083
PubChem CID 517165
ChEBI CHEBI:62947
Formula Weight 77.08g/mol
Percent Purity ≥99.99% (metals basis)
Grade TraceSELECT
Chemical Name or Material Ammonium acetate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.