missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Aluminum Sulfate Dodecahydrate, Reagent Grade, ≥99% (Titration), Solstice

Product Code. 15607640
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15607640 500 g Poly Bottle 500g
15697630 1 kg Poly Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15607640 Supplier Fluka Supplier No. A2140500G
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
This item is not returnable. View return policy

Reagent Grade,≥99% (titration)

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-26-1
Molecular Formula AlH28NO20S2
Molecular Weight (g/mol) 453.313
MDL Number MFCD00149958
InChI Key WZUKKIPWIPZMAS-UHFFFAOYSA-K
Synonym ammonia alum, unii-5c36drl9zn, aluminum ammonium sulfate dodecahydrate, aluminum ammonium disulfate dodecahydrate, ammonium aluminum sulfate hydrate, alum, ammonium usp, aluminum ammonium sulfate dodecahydrate, ammonium aluminum sulfate dodecahydrate, 5c36drl9zn, ammonium aluminum disulfate dodecahydrate
PubChem CID 62668
IUPAC Name aluminum;azanium;disulfate;dodecahydrate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3]

Specifications

Melting Point 93.5°C
Color White
pH 2.6 (at 25°C)
Quantity 500 g
Linear Formula AlNH4(SO4)2 · 12H2O
UN Number NONH for all modes of transport
Formula Weight 453.33g/mol
Percent Purity ≥99% (titration)
Odor Odorless
Packaging Poly Bottle
Appearance Crystalline
Density 1.640g/cm3 at 20°C
Chemical Name or Material Ammonium aluminum sulfate dodecahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.