missing translation for 'onlineSavingsMsg'
Learn More

Ammonium aluminum sulfate dodecahydrate, Reagent Grade, ≥99% (titration), Solstice

Product Code. 16625933
Change view
Click to view available options
10 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16625933 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16625933 Supplier Fluka Supplier No. A214010KG
  • ONLINE EXCLUSIVE PRICE
    2705.00 €
    2543.37 € / 10kg
    Save 161.63 €
    5% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 93.5°C
CAS 7784-26-1
Color White
CAS Max % 1
pH 2.6
Quantity 10 kg
Molecular Formula AlH28NO20S2
Linear Formula AlNH4(SO4)2 · 12H2O
Synonym Aluminum ammonium sulfate dodecahydrate; Ammonium alum
InChI Key WZUKKIPWIPZMAS-UHFFFAOYSA-K
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
IUPAC Name aluminium(3+) ammonium dodecahydrate disulfate
Molecular Weight (g/mol) 453.31
Grade Reagent
Density 1.640 g/cm3 (20°C)
Chemical Name or Material Ammonium aluminum sulfate dodecahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.