missing translation for 'onlineSavingsMsg'
Learn More

Ammonium bicarbonate, 98%, pure

Product Code. 10262382 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic Drum
Unit Size:
1kg
25g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10262382 1 kg Glass bottle 1kg
10364072 25 g Glass bottle 25g
11904321 5 kg Plastic Drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10262382 Supplier Thermo Scientific Acros Supplier No. 370930010
  • ONLINE EXCLUSIVE PRICE
    73.15 €
    71.00 € / 1kg
    Save 2.15 €
    2% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1066-33-7 | C3H8N2O9Zn2 | 346.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1066-33-7
Molecular Formula CH5NO3
Molecular Weight (g/mol) 79.06
MDL Number MFCD00012138
InChI Key ATRRKUHOCOJYRX-UHFFFAOYSA-N
Synonym ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327
PubChem CID 14013
IUPAC Name carbonic acid amine
SMILES N.OC(O)=O

Specifications

Melting Point 105°C
Color White
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 1 kg
Assay Percent Range 98%
Linear Formula NH4HCO3
Merck Index 15,494
Solubility Information Solubility in water: 220g/L (20°C)
Formula Weight 79.06
Percent Purity 98%
Grade Pure
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Ammonium bicarbonate
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium bicarbonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.