missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Bicarbonate, Fisher BioReagents

Product Code. 10532775
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10532775 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10532775 Supplier Fisher BioReagents Supplier No. BP2413500
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1066-33-7 | C3H8N2O9Zn2 | 346.86 g/mol

Poly Bottle; 500g

Ammonium Bicarbonate is used for many biochemical applications.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ammonium Bicarbonate
Melting Point 105°C
CAS 1066-33-7
Color White
pH 8
Physical Form Powder/Solid
Quantity 500 g
Assay Percent Range 21.30 to 21.73 %
Molecular Formula CH5NO3
Linear Formula NH4HCO3
MDL Number MFCD00012138
Merck Index 15, 494
Synonym ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327
InChI Key ATRRKUHOCOJYRX-UHFFFAOYSA-N
SMILES N.OC(O)=O
IUPAC Name carbonic acid amine
Molecular Weight (g/mol) 79.06
PubChem CID 14013
Formula Weight 79.06
Percent Purity 21.30 to 21.73%
Identification Pass Test
Packaging Poly Bottle
Vapor Pressure 67 hPa at 20°C
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium bicarbonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.