missing translation for 'onlineSavingsMsg'
Learn More

Ammonium bicarbonate, Puriss., meets analytical spec. of Ph.Eur., BP, E 503, 99-101%, Solstice
BIOPHARMA

Product Code. 15681030
Change view
Click to view available options
Quantity:
1 kg
25 kg
6 x 1 kg
Packaging:
Fiberboard Box
Poly Bottle
Unit Size:
1kg
25kg
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15681030 6 x 1 kg Poly Bottle Pack of 6
15661030 1 kg Poly Bottle 1kg
15671030 25 kg Fiberboard Box 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15681030 Supplier Honeywell-Fluka Supplier No. 112136X1KG
  • ONLINE EXCLUSIVE PRICE
    171.00 €
    154.23 € / Pack of 6
    Save 16.77 €
    9% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss., meets analytical specification of Ph.Eur., BP, E 503, 99-101%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1066-33-7
Molecular Formula CH5NO3
Molecular Weight (g/mol) 79.06
MDL Number MFCD00012138
InChI Key ATRRKUHOCOJYRX-UHFFFAOYSA-N
Synonym ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327
PubChem CID 14013
IUPAC Name carbonic acid amine
SMILES N.OC(O)=O

Specifications

CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Physical Form Powder
Quantity 6 x 1 kg
Assay Percent Range 99 to 101%
Linear Formula NH4HCO3
Beilstein 4329606
Formula Weight 79.06
Percent Purity 99 to 101%
Grade puriss.
Packaging Poly Bottle
Density 1.58 g/cm3; Vapor Density: 2.7 (vs air)
Chemical Name or Material Ammonium bicarbonate, Puriss., meets analytical specification of Ph.Eur., BP, E 503, 99-101%
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium bicarbonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Ammonium hydrogencarbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.