missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Bisulfite - In water, TRC

Product Code. 30330936
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30330936 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330936 Supplier Toronto Research Chemicals Supplier No. TRCA6341431ML
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ammonium Bisulfite
CAS 10192-30-0
Molecular Formula H5NO3S
Quantity 1 mL
Synonym Ammonium Acid Sulfite, Ammonium Bisulfite, Ammonium Bisulfite (NH4HSO3), Ammonium Hydrogen Sulfite, Ammonium Monosulfite, Ammonium Sulfite (NH4HSO3), Monoammonium Sulfite, Solfoliquid 25, TC 1000
InChI Formula InChI=1S/H3N.H2O3S/c;1-4(2)3/h1H3;(H2,1,2,3)
SMILES [NH4+].OS(=O)[O-]
IUPAC Name azanium;hydrogen sulfite
Molecular Weight (g/mol) 99.11
Formula Weight 98.999
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.