missing translation for 'onlineSavingsMsg'
Learn More

Ammonium bromide, 99%

Product Code. 11417727
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Unit Size:
10000g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11417727 500 g 500g
11427727 2500 g 2500g
11437727 10,000 g 10000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11417727 Supplier Thermo Scientific Alfa Aesar Supplier No. A16258.36
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 12124-97-9 | BrH4N | 97.94 g/mol

Ammonium bromide is often used as an intermediate source of bromide ion in laboratory processes and pharmaceutical preparations. It is most commonly used as an additive in the preparation of photographic films, fireproofing products and corrosion inhibitors. Ammonium bromide is widely utilized in the manufacture of photographic chemicals and emulsions. It is also used in pharmaceutical preparations. It finds application in fireproofing of wood and lithography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium bromide is often used as an intermediate source of bromide ion in laboratory processes and pharmaceutical preparations. It is most commonly used as an additive in the preparation of photographic films, fireproofing products and corrosion inhibitors. Ammonium bromide is widely utilized in the manufacture of photographic chemicals and emulsions. It is also used in pharmaceutical preparations. It finds application in fireproofing of wood and lithography.

Solubility
Soluble in water, methanol, ethanol and acetone. Slightly soluble in ether.

Notes
Hygroscopic and air sensitive. Incompatible with strong oxidizing agents, acids, bases, heavy metals, silver salts and potassium.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12124-97-9
Molecular Formula BrH4N
Molecular Weight (g/mol) 97.94
MDL Number MFCD00011419
InChI Key SWLVFNYSXGMGBS-UHFFFAOYSA-N
IUPAC Name ammonium bromide
SMILES [NH4+].[Br-]

Specifications

Melting Point 452°C (sublimation)
Quantity 500 g
Merck Index 14,505
Solubility Information Soluble in water,methanol,ethanol and acetone. Slightly soluble in ether.
Formula Weight 97.97
Percent Purity 99%
Sensitivity Hygroscopic
Density 2.429
Chemical Name or Material Ammonium bromide
compliance-icons
Product Identifier
  • Ammonium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Ammonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.