missing translation for 'onlineSavingsMsg'
Learn More

Ammonium bromide, Puratronic™, 99.999% (metals basis)

Product Code. 11342078
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11342078 100 g 100g
11322078 5 g 5g
11332078 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11342078 Supplier Thermo Scientific Alfa Aesar Supplier No. 012981.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 12124-97-9 | BrH4N | 97.94 g/mol

Ammonium bromide is used in pharmaceuticals for its bromide ion. It is used for a variety of analytical chemistry applications. It is also used for photography in films, plates and papers; in fireproofing of wood; in lithography and process engraving and as corrosion inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12124-97-9
Molecular Formula BrH4N
Molecular Weight (g/mol) 97.94
MDL Number MFCD00011419
InChI Key SWLVFNYSXGMGBS-UHFFFAOYSA-N
IUPAC Name ammonium bromide
SMILES [NH4+].[Br-]

Specifications

Melting Point 452°C (sublimation)
Color White
Physical Form Crystalline
Quantity 100 g
Merck Index 14,505
Solubility Information Soluble in water (970mg/ml at 25°C),methanol,ethanol,alcohol (freely),and acetone (freely).
Formula Weight 97.97
Percent Purity 99.999%
Grade Puratronic
Sensitivity Hygroscopic
Density 2.429 g/mL
Assay (metals basis)
Chemical Name or Material Ammonium bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.