missing translation for 'onlineSavingsMsg'
Learn More

Ammonium carbonate, for analysis

Product Code. 10511071 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
2.5kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10511071 1 kg Plastic bottle 1kg
10511272 100 g Plastic bottle 100g
10605652 500 g Plastic bottle 500g
10257403 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10511071 Supplier Thermo Scientific Acros Supplier No. 196650010
  • ONLINE EXCLUSIVE PRICE
    88.20 €
    85.60 € / 1kg
    Save 2.60 €
    2% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 506-87-6 | CH8N2O3 | 96.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-87-6
Molecular Formula CH8N2O3
Molecular Weight (g/mol) 96.09
MDL Number MFCD00010890
InChI Key PRKQVKDSMLBJBJ-UHFFFAOYSA-N
Synonym ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate
PubChem CID 517111
IUPAC Name diazanium;carbonate
SMILES C(=O)([O-])[O-].[NH4+].[NH4+]

Specifications

Melting Point 58.0°C
Color White
Physical Form Crystalline Powder
Quantity 1 kg
Linear Formula (NH4)2CO3
Merck Index 15, 505
Solubility Information Solubility in water: soluble (320g/L at 20°C)
Formula Weight 96.09
Percent Purity ≥30.0% (NH3)
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Ammonium carbonate, p.a.
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium carbonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Carbonic acid, diammonium salt 5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.